ContactUsLinkComponent
Biotinylated Human CD64, His,Avitag (SPR  BLI verified) (FCA-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 1 6 - Pro 288 (Accession P12314-1). 1 * 25 µG
Artikelnr. ACRBFCA-H82E8-25UG
Lieferant:  ACRO Biosystems
Biotinylated Human CD64, His,Avitag (SPR  BLI verified) (FCA-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 1 6 - Pro 288 (Accession P12314-1). 1 * 25 µG
Artikelnr. ACRBFCA-H82E8-25UG
Lieferant:  ACRO Biosystems
Frequently Bought Together