Recombinat Protein,Human CD155 PVR Protein, His Tag,Human CD155, His Tag (CD5-H5223) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accession NP006496.2).This protein carries a polyhistidine tag at the C-terminus. The ca 1 * 1 mg
Artikelnr. ACRBCD5-H5223-1MG
Lieferant: ACRO Biosystems
Frequently Bought Together