ContactUsLinkComponent
Human CD27, Fc Tag (CD7-H5254) is expressed from human 293 cells (HEK293). It contains AA Thr 21 - Ile 192 (Accession NP001233). 1 * 200 µG
Artikelnr. ACRBCD7-H5254200UG
Lieferant:  ACRO Biosystems
Human CD27, Fc Tag (CD7-H5254) is expressed from human 293 cells (HEK293). It contains AA Thr 21 - Ile 192 (Accession NP001233). 1 * 200 µG
Artikelnr. ACRBCD7-H5254200UG
Lieferant:  ACRO Biosystems
Frequently Bought Together