ContactUsLinkComponent
Human CD30 Ligand, His Tag, low endotoxin (CDL-H524b) is expressed from human 293 cells (HEK293). It contains AA Gln 63 - Asp 234 (A ccession P32971-1). 1 * 1 mg
Catalog # ACRBCDL-H524B-1MG
Supplier:  ACRO Biosystems
Human CD30 Ligand, His Tag, low endotoxin (CDL-H524b) is expressed from human 293 cells (HEK293). It contains AA Gln 63 - Asp 234 (A ccession P32971-1). 1 * 1 mg
Catalog # ACRBCDL-H524B-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together