ContactUsLinkComponent
Recombinat Protein,Human FOLR1 Protein, Strep Tag,Human FOLR1, Strep Tag (FO1-H528b) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession NP057937.1).This protein carries a twin strep tag at the C-terminus. The prot 1 * 25 µG
Artikelnr. ACRBFO1-H528B-25UG
Lieferant:  ACRO Biosystems
Recombinat Protein,Human FOLR1 Protein, Strep Tag,Human FOLR1, Strep Tag (FO1-H528b) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession NP057937.1).This protein carries a twin strep tag at the C-terminus. The prot 1 * 25 µG
Artikelnr. ACRBFO1-H528B-25UG
Lieferant:  ACRO Biosystems
Frequently Bought Together