ContactUsLinkComponent
Human IL-2 R beta, Fc Tag (SPR verified) (ILB-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (Ac cession NP000869.1). 1 * 50 µG
Catalog # ACRBILB-H5253-50UG
Supplier:  ACRO Biosystems
Human IL-2 R beta, Fc Tag (SPR verified) (ILB-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (Ac cession NP000869.1). 1 * 50 µG
Catalog # ACRBILB-H5253-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together