ContactUsLinkComponent
Biotinylated Human Dkk-1, Fc,Avitag (DK1-H82F5) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accessi on O94907-1). 1 * 25 µG
Artikelnr. ACRBDK1-H82F5-25UG
Lieferant:  ACRO Biosystems
Biotinylated Human Dkk-1, Fc,Avitag (DK1-H82F5) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accessi on O94907-1). 1 * 25 µG
Artikelnr. ACRBDK1-H82F5-25UG
Lieferant:  ACRO Biosystems
Frequently Bought Together