ContactUsLinkComponent
Human GITR Ligand, His Tag (GIL-H5249) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH69 319.1). 1 * 1 mg
Catalog # ACRBGIL-H5249-1MG
Supplier:  ACRO Biosystems
Human GITR Ligand, His Tag (GIL-H5249) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH69 319.1). 1 * 1 mg
Catalog # ACRBGIL-H5249-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together