Mouse ICOS (C137S, C138S), Fc Tag (ICS-M5259) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Leu 144 (Accession Q9WVS0-1 (C137S, C138S)). 1 * 100 µG
Artikelnr. ACRBICS-M5259100UG
Lieferant: ACROBIOSYSTEMS
Frequently Bought Together