ContactUsLinkComponent
Human PD-L1, Fc Tag (HPLC-verified) (PD1-H5258) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accessi on NP054862.1). 1 * 100 µG
Artikelnr. ACRBPD1-H5258100UG
Lieferant:  ACRO Biosystems
Human PD-L1, Fc Tag (HPLC-verified) (PD1-H5258) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accessi on NP054862.1). 1 * 100 µG
Artikelnr. ACRBPD1-H5258100UG
Lieferant:  ACRO Biosystems
Frequently Bought Together