ContactUsLinkComponent
Biotinylated Human IL17A, His,Avitag (ILA-H82Q1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (Access ion Q16552-1). 1 * 200 µG
  ACRBILA-H82Q1200UG
Biotinylated Human IL17A, His,Avitag (ILA-H82Q1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (Access ion Q16552-1). 1 * 200 µG
  ACRBILA-H82Q1200UG
Frequently Bought Together