ContactUsLinkComponent
Human Siglec-9, His Tag (SI9-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession Q9Y336-1 ). 1 * 100 µG
Artikelnr. ACRBSI9-H52H4100UG
Lieferant:  ACRO Biosystems
Human Siglec-9, His Tag (SI9-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession Q9Y336-1 ). 1 * 100 µG
Artikelnr. ACRBSI9-H52H4100UG
Lieferant:  ACRO Biosystems
Frequently Bought Together