Human LRP-6 (20-630), Mouse IgG2a Fc Tag (LR6-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 630 (Ac cession O75581-1 ). 1 * 100 µG
Artikelnr. ACRBLR6-H5253100UG
Lieferant: ACRO Biosystems
Frequently Bought Together