Recombinat Protein,Human Frizzled-7 FZD7 Protein, His Tag,Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells ( HEK293). It contains AA Gln 33 - Leu 185 (Accession O75084-1).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg
Artikelnr. ACRBFZ7-H52H9-1MG
Lieferant: ACRO Biosystems
Frequently Bought Together