ContactUsLinkComponent
Human PD-L1, Mouse IgG1 Fc Tag, low endotoxin (HPLC-verified) (PD1-H52A3) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession NP054862.1). 1 * 100 µG
Catalog # ACRBPD1-H52A3100UG
Supplier:  ACRO Biosystems
Human PD-L1, Mouse IgG1 Fc Tag, low endotoxin (HPLC-verified) (PD1-H52A3) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession NP054862.1). 1 * 100 µG
Catalog # ACRBPD1-H52A3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together