ContactUsLinkComponent
Rhesus macaque OX40, His Tag (OX0-C5220) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Ala 214 (Accession XP0 01090870.1). 1 * 1 mg
Artikelnr. ACRBOX0-C5220-1MG
Lieferant:  ACRO Biosystems
Rhesus macaque OX40, His Tag (OX0-C5220) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Ala 214 (Accession XP0 01090870.1). 1 * 1 mg
Artikelnr. ACRBOX0-C5220-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together