Recombinat Protein,Rhesus macaque SLAMF7 CRACC CD319 Protein, His Tag,Rhesus macaque SLAMF7, His Tag (SL7-R52H7) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession F7HQ72-1).This protein carries a polyhistidine t e 1 * 1 mg
Artikelnr. ACRBSL7-R52H7-1MG
Lieferant: ACRO Biosystems
Frequently Bought Together