ContactUsLinkComponent
Recombinat Protein,Human TIMP-2 Protein, His Tag,Human TIMP-2, His Tag (TI2-H5223) is expressed from human 293 cells (HEK293). It co ntains AA Cys 27 - Pro 220 (Accession NP003246.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 50 µG
  ACRBTI2-H5223-50UG
Recombinat Protein,Human TIMP-2 Protein, His Tag,Human TIMP-2, His Tag (TI2-H5223) is expressed from human 293 cells (HEK293). It co ntains AA Cys 27 - Pro 220 (Accession NP003246.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 50 µG
  ACRBTI2-H5223-50UG
Frequently Bought Together