Express Eingabe
Austria
ContactUsLinkComponent
Human FOLR1, Fc Tag (FO1-H5253) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession P15328-1). 1 * 1 mg
Catalog # ACRBFO1-H5253-1MG
Supplier:  ACRO Biosystems
Human FOLR1, Fc Tag (FO1-H5253) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession P15328-1). 1 * 1 mg
Catalog # ACRBFO1-H5253-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together