Biotinylated Human CA125, Fc,Avitag (CA5-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gly 12660 - Met 12923 (Ac cession Q8WXI7-1). 1 * 25 µG
Artikelnr. ACRBCA5-H82F4-25UG
Lieferant: ACRO Biosystems
Frequently Bought Together