ContactUsLinkComponent
Cynomolgus Glypican 3, Fc Tag (GP3-C5256) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession XP 005594665.1). 1 * 100 µG
Artikelnr. ACRBGP3-C5256100UG
Lieferant:  ACRO Biosystems
Cynomolgus Glypican 3, Fc Tag (GP3-C5256) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession XP 005594665.1). 1 * 100 µG
Artikelnr. ACRBGP3-C5256100UG
Lieferant:  ACRO Biosystems
Frequently Bought Together