ContactUsLinkComponent
Biotinylated Human CD155, Fc,Avitag (CD5-H82F6) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accessi on NP006496.2). 1 * 200 µG
Artikelnr. ACRBCD5-H82F6200UG
Lieferant:  ACRO Biosystems
Biotinylated Human CD155, Fc,Avitag (CD5-H82F6) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accessi on NP006496.2). 1 * 200 µG
Artikelnr. ACRBCD5-H82F6200UG
Lieferant:  ACRO Biosystems
Frequently Bought Together