ContactUsLinkComponent
Recombinat Protein,Human LBP Lipopolysaccharide Protein, His Tag,Human Lipopolysaccharide, His Tag (LBP-H52H5) is expressed from hu man 293 cells (HEK293). It contains AA Ala 26 - Val 481 (Accession P18428).This protein carries a polyhistidine tag a 1 * 100 µG
Artikelnr. ACRBLBP-H52H5100UG
Lieferant:  ACRO Biosystems
Recombinat Protein,Human LBP Lipopolysaccharide Protein, His Tag,Human Lipopolysaccharide, His Tag (LBP-H52H5) is expressed from hu man 293 cells (HEK293). It contains AA Ala 26 - Val 481 (Accession P18428).This protein carries a polyhistidine tag a 1 * 100 µG
Artikelnr. ACRBLBP-H52H5100UG
Lieferant:  ACRO Biosystems
Frequently Bought Together