PE-Labeled Human SIRP alpha, Fc Tag (recommended for neutralizing assay) (Cat. No. SIA-HP252) is produced via conjugation of PE to H uman SIRP alpha, Fc tag under optimal conditions. Human SIRP alpha, Fc tag is expressed in HEK293 cells containing AA 1 * 50 Tests
ACRBSIAHP25250TEST
Frequently Bought Together