ContactUsLinkComponent
Human IL-21 R, Fc Tag (ILR-H5257) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession AAH04348). 1 * 1 mg
Artikelnr. ACRBILR-H5257-1MG
Lieferant:  ACRO Biosystems
Human IL-21 R, Fc Tag (ILR-H5257) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession AAH04348). 1 * 1 mg
Artikelnr. ACRBILR-H5257-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together