Recombinat Protein,Human PSMA FOLH1 Protein, His Tag,Human PSMA, His Tag (PSA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1 ).This protein carries a polyhistidine tag at the N-terminus. The p a 1 * 100 µG
Artikelnr. ACRBPSA-H52H3100UG
Lieferant: ACRO Biosystems
Frequently Bought Together