ContactUsLinkComponent
Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 1 mg
Artikelnr. ACRBC5A-M51H4-1MG
Lieferant:  ACRO Biosystems
Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 1 mg
Artikelnr. ACRBC5A-M51H4-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together