Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 1 mg
Artikelnr. ACRBC5A-M51H4-1MG
Lieferant: ACRO Biosystems
Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 1 mg
Artikelnr. ACRBC5A-M51H4-1MG
Lieferant: ACRO Biosystems
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Lieferant: VWR Chemicals
Sealing film for manual application, Parafilm® M
Lieferant: HEATHROW
Nitrilhandschuhe, Kimtech™ Purple Nitrile™
Lieferant: KIMBERLY CLARK
VWR®, Serologische Pipetten, Standard Serie
Lieferant: VWR Collection
Acetonitril ≥99,9%, HiPerSolv CHROMANORM® für die LC-MS, geeignet für UPLC/UHPLC-Instrumente
Lieferant: VWR Chemicals
VWR®, Nitrilhandschuhe
Lieferant: VWR Collection
VWR® Gesichtstücher
Lieferant: VWR Collection
pH-Indikatorstäbchen, MQuant®, Supelco®
Lieferant: Merck
Methanol ≥99,8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Gradienten-Qualität für die HPLC
Lieferant: VWR Chemicals
Wasser, HiPerSolv CHROMANORM® für die HPLC
Lieferant: VWR Chemicals
VWR®, Transferpipetten, High Performance
Lieferant: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschuhe
Lieferant: VWR Collection