Recombinat Protein,Human TRAIL R4 TNFRSF10D Protein, His Tag,Human TRAIL R4, His Tag (TR4-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 56 - His 211 (Accession NP003831).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg
Catalog # ACRBTR4-H52H5-1MG
Supplier: ACRO Biosystems
Frequently Bought Together