ContactUsLinkComponent
Recombinat Protein,Biotinylated Human BTN3A2 Protein, His,Avitag™,Biotinylated Human BTN3A2, His,Avitag (BT2-H82E7) is expressed fro m human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession P78410-1).This protein carries a polyhistidin 1 * 200 µG
Artikelnr. ACRBBT2-H82E7200UG
Lieferant:  ACRO Biosystems
Recombinat Protein,Biotinylated Human BTN3A2 Protein, His,Avitag™,Biotinylated Human BTN3A2, His,Avitag (BT2-H82E7) is expressed fro m human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession P78410-1).This protein carries a polyhistidin 1 * 200 µG
Artikelnr. ACRBBT2-H82E7200UG
Lieferant:  ACRO Biosystems
Frequently Bought Together