ContactUsLinkComponent
Human Latent Activin A, His Tag (ACA-H424x) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ser 426 (Accession AAH07858.1). 1 * 100 µG
Artikelnr. ACRBACA-H424X100UG
Lieferant:  ACRO Biosystems
Human Latent Activin A, His Tag (ACA-H424x) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ser 426 (Accession AAH07858.1). 1 * 100 µG
Artikelnr. ACRBACA-H424X100UG
Lieferant:  ACRO Biosystems
Frequently Bought Together