ContactUsLinkComponent
Biotinylated Human VEGF R2, Avitag,His Tag (KDR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 ( Accession AAI31823). 1 * 200 µG
Catalog # ACRBKDR-H82E5200UG
Supplier:  ACRO Biosystems
Biotinylated Human VEGF R2, Avitag,His Tag (KDR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 ( Accession AAI31823). 1 * 200 µG
Catalog # ACRBKDR-H82E5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together