Recombinat Protein,Mouse gp130 CD130 IL-6 R beta Protein, His Tag,Mouse gp130, His Tag (ILT-M52H1) is expressed from human 293 cel ls (HEK293). It contains AA Gln 23 - Glu 617 (Accession NP034690).This protein carries a polyhistidine tag at the C-te 1 * 1 mg
Catalog # ACRBILT-M52H1-1MG
Supplier: ACRO Biosystems
Frequently Bought Together