0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Human TIM-3, Fc Tag (TM3-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession Q8TDQ0-1). 1 * 1 mg
ACRBTM3-H5258-1MG
:
ACRO Biosystems
Human TIM-3, Fc Tag (TM3-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession Q8TDQ0-1). 1 * 1 mg
ACRBTM3-H5258-1MG
:
ACRO Biosystems
Frequently Bought Together
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
pH-Indikatorstäbchen, MQuant®, Supelco®
: Merck
Acetonitril ≥99,9%, HiPerSolv CHROMANORM® für die LC-MS, geeignet für UPLC/UHPLC-Instrumente
: VWR Chemicals
VWR®, Nitrilhandschuhe
: VWR Collection
VWR® Gesichtstücher
: VWR Collection
Methanol ≥99,8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Gradienten-Qualität für die HPLC
: VWR Chemicals
Wasser, HiPerSolv CHROMANORM® für die HPLC
: VWR Chemicals
Acetonitril ≥99,9%, LiChrosolv® Reag. Ph. Eur., Gradienten-Qualität für die Flüssigchromatographie, Supelco®
: Merck
VWR®, Serologische Pipetten, Standard Serie
: VWR Collection
Nitrilhandschuhe, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Transferpipetten, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschuhe
: VWR Collection
Acetonitril, wasserfrei (max. 0,003% H₂O) ≥99,95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, Supergradienten-Qualität geeignet für UPLC/UHPLC-Instrumente
: VWR Chemicals