Biotinylated Human IL-15 R alpha, Fc,Avitag (ILA-H82F4) is expressed from human 293 cells (HEK293). It contains AA Ile 31 - Thr 205 (Accession Q13261-1). 1 * 200 µG
Artikelnr. ACRBILA-H82F4200UG
Lieferant: ACRO Biosystems
Frequently Bought Together