Recombinat Protein,Human LAMP1 CD107a Protein, His Tag,Human LAMP1, His Tag (LA1-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Met 382 (Accession NP005552).This protein carries a polyhistidine tag at the C-terminus. The n 1 * 1 mg
Artikelnr. ACRBLA1-H5221-1MG
Lieferant: ACRO Biosystems
Frequently Bought Together