Human PD-1, Llama IgG2b Fc Tag, low endotoxin (PD1-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 16 7 (Accession Q15116-1). 1 * 1 mg
Catalog # ACRBPD1-H5259-1MG
Supplier: ACRO Biosystems
Frequently Bought Together