ContactUsLinkComponent
Human ROR1 (39-151, Ig-like domain), His Tag (RO1-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 39 - Gly 151 (Accession Q01973-1). 1 * 1 mg
Artikelnr. ACRBRO1-H5221-1MG
Lieferant:  ACRO Biosystems
Human ROR1 (39-151, Ig-like domain), His Tag (RO1-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 39 - Gly 151 (Accession Q01973-1). 1 * 1 mg
Artikelnr. ACRBRO1-H5221-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together