ContactUsLinkComponent
Biotinylated Human TNFSF11, Avitag,Fc Tag (RAL-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (A ccession AAC51762.1). 1 * 25 µG
  ACRBRAL-H82F9-25UG
Biotinylated Human TNFSF11, Avitag,Fc Tag (RAL-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (A ccession AAC51762.1). 1 * 25 µG
  ACRBRAL-H82F9-25UG
Frequently Bought Together