ContactUsLinkComponent
Recombinat Protein,Human Glutaredoxin 1 GLRX1 Protein, His Tag,Human Glutaredoxin 1, His Tag (GLX-H5149) is expressed from E.coli c ells. It contains AA Met 1 - Gln 106 (Accession NP001112362).This protein carries a polyhistidine tag at the N-termin 1 * 1 mg
Artikelnr. ACRBGLX-H5149-1MG
Lieferant:  ACRO Biosystems
Recombinat Protein,Human Glutaredoxin 1 GLRX1 Protein, His Tag,Human Glutaredoxin 1, His Tag (GLX-H5149) is expressed from E.coli c ells. It contains AA Met 1 - Gln 106 (Accession NP001112362).This protein carries a polyhistidine tag at the N-termin 1 * 1 mg
Artikelnr. ACRBGLX-H5149-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together