ContactUsLinkComponent
Biotinylated Human Siglec-15, Fc,Avitag (SG5-H82F5) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Acc ession Q6ZMC9-1). 1 * 25 µG
Artikelnr. ACRBSG5-H82F5-25UG
Lieferant:  ACRO Biosystems
Biotinylated Human Siglec-15, Fc,Avitag (SG5-H82F5) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Acc ession Q6ZMC9-1). 1 * 25 µG
Artikelnr. ACRBSG5-H82F5-25UG
Lieferant:  ACRO Biosystems
Frequently Bought Together