ContactUsLinkComponent
Biotinylated Human VSIG3, Fc,Avitag (VS3-H82F9) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 241 (Accessi on Q5DX21-1). 1 * 25 µG
Artikelnr. ACRBVS3-H82F9-25UG
Lieferant:  ACRO Biosystems
Biotinylated Human VSIG3, Fc,Avitag (VS3-H82F9) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 241 (Accessi on Q5DX21-1). 1 * 25 µG
Artikelnr. ACRBVS3-H82F9-25UG
Lieferant:  ACRO Biosystems
Frequently Bought Together