Human CD30, Llama IgG2b Fc Tag, low endotoxin (TN8-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 37 9 (Accession NP001234.2). 1 * 1 mg
Artikelnr. ACRBTN8-H5250-1MG
Lieferant: ACRO Biosystems
Frequently Bought Together