ContactUsLinkComponent
Human TSLP, His Tag (TSP-H52Hb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession Q969D9-1). 1 * 100 µG
Artikelnr. ACRBTSP-H52HB100UG
Lieferant:  ACRO Biosystems
Human TSLP, His Tag (TSP-H52Hb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession Q969D9-1). 1 * 100 µG
Artikelnr. ACRBTSP-H52HB100UG
Lieferant:  ACRO Biosystems
Frequently Bought Together