ContactUsLinkComponent
Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, Fc Tag,Mouse TIM-3, Fc Tag (TI3-M5252) is expressed from human 293 cells (HEK293). I t contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a human IgG1 Fc tag at the C-terminus. Th a 1 * 1 mg
Artikelnr. ACRBTI3-M5252-1MG
Lieferant:  ACRO Biosystems
Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, Fc Tag,Mouse TIM-3, Fc Tag (TI3-M5252) is expressed from human 293 cells (HEK293). I t contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a human IgG1 Fc tag at the C-terminus. Th a 1 * 1 mg
Artikelnr. ACRBTI3-M5252-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together