ContactUsLinkComponent
Recombinat Protein,Human IL-18BP Protein, His Tag,Human IL-18BP, His Tag (ILP-H5222) is expressed from human 293 cells (HEK293). It contains AA Thr 31 - Gly 194 (Accession NP001138529).This protein carries a polyhistidine tag at the C-terminus. The c 1 * 200 µG
  ACRBILP-H5222200UG
Recombinat Protein,Human IL-18BP Protein, His Tag,Human IL-18BP, His Tag (ILP-H5222) is expressed from human 293 cells (HEK293). It contains AA Thr 31 - Gly 194 (Accession NP001138529).This protein carries a polyhistidine tag at the C-terminus. The c 1 * 200 µG
  ACRBILP-H5222200UG
Frequently Bought Together