ContactUsLinkComponent
Human LRP-5, Mouse IgG2a Fc Tag (LR5-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 644 - Gln 1263 (Accession O75197-1 ). 1 * 100 µG
Catalog # ACRBLR5-H5254100UG
Supplier:  ACRO Biosystems
Human LRP-5, Mouse IgG2a Fc Tag (LR5-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 644 - Gln 1263 (Accession O75197-1 ). 1 * 100 µG
Catalog # ACRBLR5-H5254100UG
Supplier:  ACRO Biosystems
Frequently Bought Together