ContactUsLinkComponent
Recombinat Protein,Human CLEC12A MICL CLL-1 Protein, His Tag,Human CLEC12A, His Tag (CLA-H5245) is expressed from human 293 cells (HEK293). It contains AA His 75 - Ala 275 (Accession NP001193939.1).This protein carries a polyhistidine tag at the N-t 1 * 1 mg
Artikelnr. ACRBCLA-H5245-1MG
Lieferant:  ACRO Biosystems
Recombinat Protein,Human CLEC12A MICL CLL-1 Protein, His Tag,Human CLEC12A, His Tag (CLA-H5245) is expressed from human 293 cells (HEK293). It contains AA His 75 - Ala 275 (Accession NP001193939.1).This protein carries a polyhistidine tag at the N-t 1 * 1 mg
Artikelnr. ACRBCLA-H5245-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together