ContactUsLinkComponent
Human VLDL R, His Tag (VLR-H5227) is expressed from human 293 cells (HEK293). It contains AA Gly 28 - Ser 769 (Accession P98155-2). 1 * 1 mg
  ACRBVLR-H5227-1MG
Human VLDL R, His Tag (VLR-H5227) is expressed from human 293 cells (HEK293). It contains AA Gly 28 - Ser 769 (Accession P98155-2). 1 * 1 mg
  ACRBVLR-H5227-1MG
Frequently Bought Together