ContactUsLinkComponent
Biotinylated Mouse FCGRTB2M Heterodimer Protein, His,Avitag (BLI verified) (FCM-M82W5) is expressed from human 293 cells (HEK293). I t contains AA Ser 22 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession  Q61559-1 (FCGRT) P01887-1 (B2M)). 1 * 25 µG
  ACRBFCM-M82W5-25UG
Biotinylated Mouse FCGRTB2M Heterodimer Protein, His,Avitag (BLI verified) (FCM-M82W5) is expressed from human 293 cells (HEK293). I t contains AA Ser 22 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession  Q61559-1 (FCGRT) P01887-1 (B2M)). 1 * 25 µG
  ACRBFCM-M82W5-25UG
Frequently Bought Together