ASTM Method D3710 Quantitative Calibration Standard
ASTM Method D3710 Quantitative Calibration Standard
Artikelnr. EHERGA0900105
Anbieter: EHRENSTORFER
ASTM Method D3710 Quantitative Calibration Standard
Artikelnr. EHERGA0900105
Anbieter: EHRENSTORFER
CAS Nummer:
Springe zu:
Product Details & Documents
- SMILES: See Mixture Tab
- Inchi: See Mixture Tab
- IUPAC: See Mixture Tab
- Packing group: II
Spezifikationen
Spezifikationen
- Bezeichnung:ASTM Method D3710 Quantitative Calibration Standard
- UN No.:1993
- Cat. No.:EHERGA0900105
- GHS-Signal:Gefahr
- Beschreibung Gefahrenklasse:GHS02, GHS07, GHS08, GHS09
- Lagerbedingungen:20 °C
- Shipping conditions:20 °C
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



